7-Aminodesacetoxycephalosporanic acid 97%

Supplier: Ambeed
Danger

Synonyms: (6R,7R)-7-Amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Aminodeacetoxycephalosporanic acid, 7-Amino-3-methyl-3-cephem-4-carboxylic acid, 7-Aminodesacetoxycephalosporanic Acid, 7-ADCA

A125641-100G A125641-10G A125641-25G A125641-5G
76996-078EA 83.82 USD
76996-078 76996-080 76996-082 76996-084
7-Aminodesacetoxycephalosporanic acid 97%
7-Aminodesacetoxycephalosporanic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃S
MW: 214.24 g/mol
MDL Number: MFCD00151456
CAS Number: 22252-43-3
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR