6-Fluoronaphthalen-2-ol 95%

Supplier: Ambeed
Warning

A125623-100MG A125623-1G A125623-250MG A125623-5G
77009-620EA 35 USD
77009-620 77009-622 77009-624 77009-626
6-Fluoronaphthalen-2-ol 95%
6-Fluoronaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₇FO
MDL Number: MFCD18412124
CAS Number: 13101-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR