2-(2-Methoxyethyl)piperidine hydrochloride 95%

Supplier: Ambeed
Warning

A125615-1G A125615-250MG A125615-5G A125615-10G
76983-452EA 38 USD
76983-452 76983-454 76983-456 77889-868
2-(2-Methoxyethyl)piperidine hydrochloride 95%
2-(2-Methoxyethyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO
MDL Number: MFCD07368304
CAS Number: 1185088-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR