2,3,5-Tri-o-benzyl-1-o-p-nitrobenzoyl-D-arabinofuranose 97%

Supplier: Ambeed
Warning

Synonyms: 2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose

A125569-1G A125569-250MG A125569-25G
76996-062EA 26.53 USD
76996-062 76996-064 76996-066
2,3,5-Tri-o-benzyl-1-o-p-nitrobenzoyl-D-arabinofuranose 97%
2,3,5-Tri-o-benzyl-1-o-p-nitrobenzoyl-D-arabinofuranose

Precaución: for Research Use Only

Formula: C₃₃H₃₁NO₈
MW: 569.61 g/mol
MDL Number: MFCD00038477
CAS Number: 52522-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR