3-Benzyloxy-4-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzyloxy)-4-fluorophenylboronic acid

A125435-1G A125435-250MG A125435-5G
77026-754EA 31 USD
77026-754 77026-756 77871-564
3-Benzyloxy-4-fluorophenylboronic acid 97%
3-Benzyloxy-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD09475829
CAS Number: 957034-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR