6-Bromo-1H-indole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A125324-10G A125324-1G A125324-100MG A125324-250MG A125324-5G
77026-742EA 187.78 USD
77026-742 77026-744 77571-512 77670-772 77670-774
6-Bromo-1H-indole-3-carboxylic acid 98%
6-Bromo-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.0500 g/mol
MDL Number: MFCD05664008
CAS Number: 101774-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR