4-Bromo-5-fluoro-2-iodoaniline 95%
Supplier: Ambeed
|
Warning
|
A125015-1G
A125015-250MG
A125015-5G
77009-612EA
80.63
USD
77009-612
77009-614
77683-112
4-Bromo-5-fluoro-2-iodoaniline 95%
4-Bromo-5-fluoro-2-iodoaniline
Precaución: For research use only.
Formula:
C₆H₄BrFIN MW: 315.91 g/mol |
MDL Number:
MFCD26386356 CAS Number: 1219741-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...