2,4-Dihydroxy-5-methoxypyrimidine 96%

Supplier: Ambeed
Warning

A124929-100G A124929-10G A124929-1G A124929-25G A124929-500G A124929-5G
77009-594EA 41 USD
77009-594 77009-596 77009-598 77009-600 77009-602 77009-604
2,4-Dihydroxy-5-methoxypyrimidine 96%
2,4-Dihydroxy-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MDL Number: MFCD00187903
CAS Number: 6623-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR