(1S,5R)-3-tert-Butoxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: rel-(1R,5S,6r)-3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

A124863-100MG A124863-10G A124863-1G A124863-250MG A124863-5G A124863-25G
77026-722EA 9 USD
77026-722 77026-724 77026-726 77026-728 77026-730 77871-562
(1S,5R)-3-tert-Butoxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid 95%
(1S,5R)-3-tert-Butoxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD12198680
CAS Number: 927679-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR