4-Chloro-1H-pyrazole-5-carboxylic acid 96%

Supplier: Ambeed
Warning

A124760-100MG A124760-1G A124760-250MG
77026-716EA 16.98 USD
77026-716 77026-718 77026-720
4-Chloro-1H-pyrazole-5-carboxylic acid 96%
4-Chloro-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃ClN₂O₂
MW: 146.532 g/mol
MDL Number: MFCD00466338
CAS Number: 84547-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR