4-Bromo-2-phenyloxazole 97%

Supplier: Ambeed
Warning

A124722-100MG A124722-1G A124722-250MG
76983-420EA 35 USD
76983-420 76983-422 77885-111
4-Bromo-2-phenyloxazole 97%
4-Bromo-2-phenyloxazole

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD12828098
CAS Number: 861440-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR