3-Bromo-6-chloro-1H-pyrazolo[3,4-d]pyrimidine 95%

Supplier: Ambeed
Warning

A124641-100MG A124641-1MG
77026-712EA 112.46 USD
77026-712 77026-714
3-Bromo-6-chloro-1H-pyrazolo[3,4-d]pyrimidine 95%
3-Bromo-6-chloro-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₄
MW: 233.45 g/mol
MDL Number: MFCD10697158
CAS Number: 944903-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR