(R)-3-(3,4-Dihydroxyphenyl)-2-hydroxypropanoic acid 98%

Supplier: Ambeed
Warning

A124609-100MG A124609-10MG A124609-1MG
76983-414EA 116 USD
76983-414 76983-416 76983-418
(R)-3-(3,4-Dihydroxyphenyl)-2-hydroxypropanoic acid 98%
(R)-3-(3,4-Dihydroxyphenyl)-2-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₅
MDL Number: MFCD09038703
CAS Number: 76822-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR