Methyl 2-bromo-6-nitrobenzoate 98%

Supplier: Ambeed
Warning

A124596-100MG A124596-1G A124596-250MG
77009-588EA 19 USD
77009-588 77009-590 77009-592
Methyl 2-bromo-6-nitrobenzoate 98%
Methyl 2-bromo-6-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MDL Number: MFCD11977361
CAS Number: 135484-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR