1-Boc-3-benzylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Benzylpiperazine

A124543-100MG A124543-10G A124543-1G A124543-250MG A124543-25G A124543-5G
77026-700EA 22.28 USD
77026-700 77026-702 77026-704 77026-706 77026-708 77026-710
1-Boc-3-benzylpiperazine 97%
1-Boc-3-benzylpiperazine

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
MDL Number: MFCD07367736
CAS Number: 502649-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR