(S)-tert-Butyl 5-bromo-2-((tert-butoxycarbonyl)amino)pentanoate 95%

Supplier: Ambeed
Warning

A124533-100MG A124533-1G A124533-250MG A124533-5G
76983-404EA 82.76 USD
76983-404 76983-406 76983-408 77592-420
(S)-tert-Butyl 5-bromo-2-((tert-butoxycarbonyl)amino)pentanoate 95%
(S)-tert-Butyl 5-bromo-2-((tert-butoxycarbonyl)amino)pentanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₆BrNO₄
MW: 352.26 g/mol
MDL Number: MFCD22576481
CAS Number: 91229-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR