4,5-Dichloropyridin-2(1H)-one 96%

Supplier: Ambeed
Warning

A124529-100MG A124529-250MG
76983-402EA 76 USD
76983-402 77889-860
4,5-Dichloropyridin-2(1H)-one 96%
4,5-Dichloropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃Cl₂NO
MDL Number: MFCD10574711
CAS Number: 856965-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR