4-(1H-Imidazol-2-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A124521-100MG A124521-10G A124521-1G A124521-250MG A124521-5G A124521-25G
76983-392EA 6 USD
76983-392 76983-394 76983-396 76983-398 76983-400 77889-859
4-(1H-Imidazol-2-yl)benzoic acid 97%
4-(1H-Imidazol-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00456057
CAS Number: 108035-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR