Morpholine-3-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A124479-100MG
A124479-1G
A124479-250MG
77009-574EA
12.74
USD
77009-574
77009-576
77009-578
Morpholine-3-carboxylic acid hydrochloride 95%
Morpholine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₀ClNO₃ MW: 167.59 g/mol |
MDL Number:
MFCD08752601 CAS Number: 66937-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...