2-(2,6-Dimethylphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A124445-1G A124445-250MG A124445-5G
77026-688EA 122.01 USD
77026-688 77026-690 77026-692
2-(2,6-Dimethylphenyl)acetic acid 98%
2-(2,6-Dimethylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
MDL Number: MFCD06208111
CAS Number: 938-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR