2,3-Dihydroxypropyl 2-(((7-chloroquinolin-4-yl)amino)benzoate 98%

Supplier: Ambeed
Warning

A124438-100MG A124438-1G A124438-1MG A124438-250MG A124438-5G
76983-382EA 16.98 USD
76983-382 76983-384 76983-386 76983-388 76983-390
2,3-Dihydroxypropyl 2-(((7-chloroquinolin-4-yl)amino)benzoate 98%
2,3-Dihydroxypropyl 2-(((7-chloroquinolin-4-yl)amino)benzoate

Precaución: for Research Use Only

Formula: C₁₉H₁₇ClN₂O₄
MW: 372.80 g/mol
MDL Number: MFCD00079326
CAS Number: 3820-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR