1-(2-Methoxyphenyl)-N-methylmethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A124241-10G A124241-5G
77026-684EA 228 USD
77026-684 77026-686
1-(2-Methoxyphenyl)-N-methylmethanamine hydrochloride 97%
1-(2-Methoxyphenyl)-N-methylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO
MDL Number: MFCD07106792
CAS Number: 181880-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR