4-Trifluoromethyl-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: H-Phe(4-CF3)-OH, 4-(Trifluoromethyl)-L-phenylalanine

A124181-10G A124181-1G A124181-250MG A124181-25G A124181-5G
76995-992EA 167 USD
76995-992 76995-994 76995-996 76995-998 76996-000
4-Trifluoromethyl-L-phenylalanine 97%
4-Trifluoromethyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
MDL Number: MFCD00044946
CAS Number: 114926-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR