7-Methyl-1,2,3,4-tetrahydroisoquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 7-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

A124139-1G A124139-250MG A124139-5G
77009-560EA 140.04 USD
77009-560 77009-562 77009-564
7-Methyl-1,2,3,4-tetrahydroisoquinoline 98%
7-Methyl-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.2200 g/mol
MDL Number: MFCD03004622
CAS Number: 207451-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR