(2,3-Difluoro-4-hydroxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A123982-100MG A123982-1G A123982-250MG A123982-5G
77026-668EA 10 USD
77026-668 77026-670 77026-672 77026-674
(2,3-Difluoro-4-hydroxyphenyl)boronic acid 97%
(2,3-Difluoro-4-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₃
MDL Number: MFCD09996974
CAS Number: 1261169-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR