3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Hydroxy-2-methoxyphenylboronic acid pinacol ester
A123909-100MG
A123909-250MG
A123909-1G
76983-366EA
45.62
USD
76983-366
76983-368
77683-105
3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Precaución: For research use only.
Formula:
C₁₃H₁₉BO₄ MW: 250.09 g/mol |
MDL Number:
MFCD16994337 CAS Number: 507462-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...