2-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
A123894-10G
A123894-1G
A123894-250MG
77026-658EA
149
USD
77026-658
77026-660
77026-662
2-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)acetic acid 97%
2-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₆ClNO₄ |
MDL Number:
MFCD03426366 CAS Number: 209525-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...