(S)-2-N-Boc-propane-1,2-diamine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 1-aminopropan-2-ylcarbamate

A123889-100MG A123889-1G A123889-250MG
76995-964EA 17 USD
76995-964 76995-966 76995-968
(S)-2-N-Boc-propane-1,2-diamine 95%
(S)-2-N-Boc-propane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD08726031
CAS Number: 146552-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR