3-(Methoxymethyl)azetidine hydrochloride 95%

Supplier: Ambeed
Warning

A123787-100MG A123787-1G
77026-656EA 21 USD
77026-656 77566-368
3-(Methoxymethyl)azetidine hydrochloride 95%
3-(Methoxymethyl)azetidine hydrochloride

Precaución: For research use only.

Formula: C₅H₁₂ClNO
MDL Number: MFCD09743447
CAS Number: 942308-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR