2-Amino-5-bromo-4-phenylpyrimidine 97%

Supplier: Ambeed
Warning

A123670-100MG A123670-10G A123670-1G A123670-250MG A123670-5G
77026-644EA 110 USD
77026-644 77026-646 77026-648 77026-650 77026-652 77026-654
2-Amino-5-bromo-4-phenylpyrimidine 97%
2-Amino-5-bromo-4-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN₃
MDL Number: MFCD09999169
CAS Number: 85658-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR