Phenyl(pyridin-2-yl)methanamine 95%

Supplier: Ambeed
Danger

A123612-100MG A123612-250MG A123612-1G
77026-630EA 11.67 USD
77026-630 77026-632 77571-436
Phenyl(pyridin-2-yl)methanamine 95%
Phenyl(pyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C12H12N2
MW: 184.2371 g/mol
MDL Number: MFCD00514135
CAS Number: 39930-11-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR