3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A123548-250MG A123548-1G
76995-958EA 40 USD
76995-958 77880-995
3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95%
3-(4-Fluorophenoxy)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClFNO
MDL Number: MFCD09879244
CAS Number: 23123-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR