2-Amino-6-methylpyridine 98%

Supplier: Ambeed
Danger

Synonyms: 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline

A123537-100G A123537-25G A123537-500G
77009-538EA 111.4 USD
77009-538 77009-540 77009-542 77009-544
2-Amino-6-methylpyridine 98%
2-Amino-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 214.4
MDL Number: MFCD00006331
CAS Number: 1824-81-3
UN: 2811
ADR: 6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR