Methyl 4,6-dibromopicolinate 95%

Supplier: Ambeed
Warning

A123525-100MG A123525-1G A123525-250MG
76983-348EA 93.36 USD
76983-348 76983-350 76983-352
Methyl 4,6-dibromopicolinate 95%
Methyl 4,6-dibromopicolinate

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MW: 294.928 g/mol
MDL Number: MFCD14582014
CAS Number: 1206248-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR