2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A123424-1G
A123424-250MG
A123424-100MG
A123424-5G
76983-342EA
251
USD
76983-342
76983-344
77889-854
77889-855
2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%
2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ |
MDL Number:
MFCD02258132 CAS Number: 192385-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...