Acacetin 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-one, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxy-5,7-dihydroxyflavone, 5,7-Dihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyrone, Linarigenin, Buddleoflavonol, 4'-Methoxyapigenin

A123393-10MG A123393-1G A123393-25G A123393-25MG A123393-5G A123393-5MG
77026-602EA 8.49 USD
77026-602 77026-604 77026-606 77026-608 77026-610 77026-612
Acacetin 98%
Acacetin

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
MDL Number: MFCD00016936
CAS Number: 480-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR