1-[3-(Difluoromethoxy)phenyl]ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(3-(Difluoromethoxy)phenyl)ethanone, 3'-(Difluoromethoxy)acetophenone

A123264-10G A123264-5G
77026-596EA 244.01 USD
77026-596 77026-598
1-[3-(Difluoromethoxy)phenyl]ethanone 95%
1-[3-(Difluoromethoxy)phenyl]ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD00236220
CAS Number: 101975-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR