4-Ethoxy-2,6-difluorophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Difluoro-4-ethoxyphenylboronic acid, (4-Ethoxy-2,6-difluorophenyl)boronic acid
A123200-1G
A123200-250MG
A123200-5G
76995-950EA
32
USD
76995-950
76995-952
76995-954
4-Ethoxy-2,6-difluorophenylboronic acid 97%
4-Ethoxy-2,6-difluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BF₂O₃ MW: 201.97 g/mol |
MDL Number:
MFCD12026735 CAS Number: 1310403-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...