1-(4-Fluorophenyl)pentan-1-one 98%

Supplier: Ambeed
Warning

A123081-1G A123081-5G
77009-530EA 14.86 USD
77009-530 77009-532
1-(4-Fluorophenyl)pentan-1-one 98%
1-(4-Fluorophenyl)pentan-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO
MW: 180.21 g/mol
MDL Number: MFCD00017968
CAS Number: 709-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR