4-(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalene-2-carbonyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: LG-100064

A123065-1MG A123065-25MG
77026-578EA 8.49 USD
77026-578 77026-580
4-(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalene-2-carbonyl)benzoic acid 97%
4-(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalene-2-carbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₆O₃
MW: 350.45 g/mol
MDL Number: MFCD13191327
CAS Number: 153559-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR