3-Bromo-6-chloropicolinaldehyde 97%

Supplier: Ambeed
Warning

A122936-100MG A122936-250MG A122936-1G
76983-330EA 24 USD
76983-330 77592-394 77889-850
3-Bromo-6-chloropicolinaldehyde 97%
3-Bromo-6-chloropicolinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
MDL Number: MFCD13189344
CAS Number: 1060815-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR