6-Chloropyrazin-2-ol 98%

Supplier: Ambeed
Warning

A122934-10G A122934-1G A122934-250MG A122934-25G A122934-5G
77026-568EA 86 USD
77026-568 77026-570 77026-572 77026-574 77026-576
6-Chloropyrazin-2-ol 98%
6-Chloropyrazin-2-ol

Precaución: for Research Use Only

Formula: C₄H₃ClN₂O
MDL Number: MFCD00204104
CAS Number: 4925-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR