α,α,α-Trifluoro-2,3-diaminotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diaminobenzotrifluoride, 3-(Trifluoromethyl)-1,2-phenylenediamine, 3-(Trifluoromethyl)benzene-1,2-diamine, 3-(Trifluoromethyl)-o-phenylenediamine

A122830-1G A122830-250MG
76995-932EA 99.73 USD
76995-932 76995-934
α,α,α-Trifluoro-2,3-diaminotoluene 95%
α,α,α-Trifluoro-2,3-diaminotoluene

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
MDL Number: MFCD08532495
CAS Number: 360-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR