(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, 4-Oxo-L-pipecolinic acid, N-BOC protected, (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, (S)-1-Boc-4-oxopipecolic acid, (2S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
A122674-10G
A122674-25G
A122674-5G
A122674-1G
77026-538EA
355.41
USD
77026-538
77026-540
77026-542
77666-762
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 97%
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₇NO₅ MW: 243.26 g/mol |
MDL Number:
MFCD01861764 CAS Number: 198646-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...