3-Chloro-4-morpholinobenzaldehyde 97%

Supplier: Ambeed
Warning

A122661-100MG A122661-1G A122661-250MG
77026-532EA 19.1 USD
77026-532 77026-534 77026-536
3-Chloro-4-morpholinobenzaldehyde 97%
3-Chloro-4-morpholinobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO₂
MW: 225.6700 g/mol
MDL Number: MFCD06740107
CAS Number: 886500-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR