tert-Butyl-5-amino-1H-indazole-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1H-indazole-1-carboxylic acid, 1,1 dimethyl ethyl ester, tert-Butyl 5-amino-1H-indazole-1-carboxylate, 1-Boc-5-amino-indazole, 5-Amino-indazole-1-carboxylic acid tert-butyl ester, N-(1)-Boc-5-aminoindazole

A122654-10G A122654-1G A122654-250MG A122654-5G A122654-25G
77009-496EA 84.88 USD
77009-496 77009-498 77009-500 77009-502 77683-101
tert-Butyl-5-amino-1H-indazole-1-carboxylate 97%
tert-Butyl-5-amino-1H-indazole-1-carboxylate

Precaución: For research use only.

Formula: C₁₂H₁₅N₃O₂
MW: 233.27 g/mol
MDL Number: MFCD04114656
CAS Number: 129488-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR