2,6-Dichloro-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A122501-1G A122501-250MG A122501-5G A122501-25G
77026-516EA 31.83 USD
77026-516 77026-518 77026-520 77566-310
2,6-Dichloro-4-methoxypyridine 98%
2,6-Dichloro-4-methoxypyridine

Precaución: For research use only.

Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
MDL Number: MFCD00955755
CAS Number: 17228-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR