tert-Butyl 5-hydroxy-3-iodo-1H-indazole-1-carboxylate 97%

Supplier: Ambeed
Warning

A122474-100MG A122474-1G
77009-492EA 108.22 USD
77009-492 77009-494
tert-Butyl 5-hydroxy-3-iodo-1H-indazole-1-carboxylate 97%
tert-Butyl 5-hydroxy-3-iodo-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃IN₂O₃
MW: 360.14 g/mol
MDL Number: MFCD27923516
CAS Number: 1426425-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR