3-Oxo-3-pyridin-4-ylpropanenitrile 95%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3-(pyridin-4-yl)propanenitrile

A122396-1G A122396-250MG A122396-5G
76995-916EA 65.78 USD
76995-916 76995-918 77571-362
3-Oxo-3-pyridin-4-ylpropanenitrile 95%
3-Oxo-3-pyridin-4-ylpropanenitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD06656807
CAS Number: 23821-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR