DL-α-Phenylglycine 97%

Supplier: Ambeed
Warning

Synonyms: Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine

A122323-100G A122323-25G A122323-500G A122323-1000G
76995-910EA 18 USD
76995-910 76995-912 76995-914 77599-764
DL-α-Phenylglycine 97%
DL-α-Phenylglycine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00064402
CAS Number: 2835-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR